missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxaprozin 98.0+%, TCI America™
$193.86 - $486.52
Chemical Identifiers
| CAS | 21256-18-8 |
|---|---|
| Molecular Formula | C18H15NO3 |
| Molecular Weight (g/mol) | 293.32 |
| MDL Number | MFCD00215977 |
| InChI Key | OFPXSFXSNFPTHF-UHFFFAOYSA-N |
| Synonym | oxaprozin, daypro, alvo, 3-4,5-diphenyl-1,3-oxazol-2-yl propanoic acid, oxaprozine, 3-4,5-diphenyloxazol-2-yl propanoic acid, deflam, oxaprozina, oxaprozinum, voir |
| PubChem CID | 4614 |
| ChEBI | CHEBI:7822 |
| IUPAC Name | 3-(4,5-diphenyl-1,3-oxazol-2-yl)propanoic acid |
| SMILES | OC(=O)CCC1=NC(=C(O1)C1=CC=CC=C1)C1=CC=CC=C1 |
Chemical Identifiers
| 21256-18-8 | |
| 293.32 | |
| OFPXSFXSNFPTHF-UHFFFAOYSA-N | |
| 4614 | |
| 3-(4,5-diphenyl-1,3-oxazol-2-yl)propanoic acid |
| C18H15NO3 | |
| MFCD00215977 | |
| oxaprozin, daypro, alvo, 3-4,5-diphenyl-1,3-oxazol-2-yl propanoic acid, oxaprozine, 3-4,5-diphenyloxazol-2-yl propanoic acid, deflam, oxaprozina, oxaprozinum, voir | |
| CHEBI:7822 | |
| OC(=O)CCC1=NC(=C(O1)C1=CC=CC=C1)C1=CC=CC=C1 |
Specifications
| 21256-18-8 | |
| White | |
| MFCD00215977 | |
| oxaprozin, daypro, alvo, 3-4,5-diphenyl-1,3-oxazol-2-yl propanoic acid, oxaprozine, 3-4,5-diphenyloxazol-2-yl propanoic acid, deflam, oxaprozina, oxaprozinum, voir | |
| OC(=O)CCC1=NC(=C(O1)C1=CC=CC=C1)C1=CC=CC=C1 | |
| 293.32 | |
| CHEBI:7822 | |
| ≥98.0% (HPLC,T) | |
| Oxaprozin |
| 163°C | |
| C18H15NO3 | |
| 5 g | |
| OFPXSFXSNFPTHF-UHFFFAOYSA-N | |
| 3-(4,5-diphenyl-1,3-oxazol-2-yl)propanoic acid | |
| 4614 | |
| 293.32 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : RP6990000
TSCA : No