missing translation for 'onlineSavingsMsg'
Learn More

Oxaprozin 98.0+%, TCI America™

Catalog No. O03775G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O03775G 5 g
O037725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O03775G Supplier TCI America Supplier No. O03775G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 21256-18-8
Molecular Formula C18H15NO3
Molecular Weight (g/mol) 293.32
MDL Number MFCD00215977
InChI Key OFPXSFXSNFPTHF-UHFFFAOYSA-N
Synonym oxaprozin, daypro, alvo, 3-4,5-diphenyl-1,3-oxazol-2-yl propanoic acid, oxaprozine, 3-4,5-diphenyloxazol-2-yl propanoic acid, deflam, oxaprozina, oxaprozinum, voir
PubChem CID 4614
ChEBI CHEBI:7822
IUPAC Name 3-(4,5-diphenyl-1,3-oxazol-2-yl)propanoic acid
SMILES OC(=O)CCC1=NC(=C(O1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 163°C
Color White
Quantity 5 g
Formula Weight 293.32
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Oxaprozin
RTECSNumber RP6990000
TSCA No