missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Oxaprozin
Chemical Identifiers
| CAS | 21256-18-8 |
|---|---|
| Molecular Formula | C18H15NO3 |
| Molecular Weight (g/mol) | 293.32 |
| MDL Number | MFCD00215977 |
| InChI Key | OFPXSFXSNFPTHF-UHFFFAOYSA-N |
| IUPAC Name | 3-(4,5-diphenyl-1,3-oxazol-2-yl)propanoic acid |
| SMILES | OC(=O)CCC1=NC(=C(O1)C1=CC=CC=C1)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC463530050
|
Thermo Scientific™
463530050 |
5 g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC463530250
|
Thermo Scientific™
463530250 |
25 g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 21256-18-8 | |
| 293.32 | |
| OFPXSFXSNFPTHF-UHFFFAOYSA-N | |
| OC(=O)CCC1=NC(=C(O1)C1=CC=CC=C1)C1=CC=CC=C1 |
| C18H15NO3 | |
| MFCD00215977 | |
| 3-(4,5-diphenyl-1,3-oxazol-2-yl)propanoic acid |
Specifications
| 21256-18-8 | |
| MFCD00215977 | |
| Solubility aq. soln.: insoluble. Other solubilities: soluble in alcohols, very slightly soluble in ether | |
| OC(=O)CCC1=NC(=C(O1)C1=CC=CC=C1)C1=CC=CC=C1 | |
| 293.32 | |
| Oxaprozin |
| C18H15NO3 | |
| 5 g | |
| OFPXSFXSNFPTHF-UHFFFAOYSA-N | |
| 3-(4,5-diphenyl-1,3-oxazol-2-yl)propanoic acid | |
| 293.32 |
Safety and Handling
GHS H Statement Toxic to aquatic life with long lasting effects.
GHS P Statement Avoid release to the environment.
Collect spillage.
Dispose of contents/container to an approved waste disposal plant.
GHS Signal Word: Warning
EINECSNumber : 244-296-1
RUO â Research Use Only