missing translation for 'onlineSavingsMsg'
Learn More

Oxamide 98.0+%, TCI America™

Catalog No. O008625G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O008625G 25 g
O0086100G 100 g
O0086500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. O008625G Supplier TCI America Supplier No. O008625G

Chemical Identifiers

CAS 471-46-5
Molecular Formula C2H4N2O2
Molecular Weight (g/mol) 88.066
MDL Number MFCD00008007
InChI Key YIKSCQDJHCMVMK-UHFFFAOYSA-N
Synonym ethanediamide, oxalamide, oxamimidic acid, oxalic acid diamide, diaminoglyoxal, oxamid, amid kyseliny stavelove, formimidic acid, 1-carbamoyl, oxamid czech, unii-sbe4m0223e
PubChem CID 10113
ChEBI CHEBI:48248
IUPAC Name oxamide
SMILES C(=O)(C(=O)N)N

Specifications

Color White
Quantity 25 g
Formula Weight 88.07
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Oxamide
EINECSNumber (2)-3137
RTECSNumber RO4900000
TSCA Yes