missing translation for 'onlineSavingsMsg'
Learn More

Oxamic Hydrazide 95.0+%, TCI America™

Catalog No. O01565G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O01565G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O01565G Supplier TCI America Supplier No. O01565G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 515-96-8
Molecular Formula C2H5N3O2
Molecular Weight (g/mol) 103.081
MDL Number MFCD00008004
InChI Key MOKRDWKSHLLYKM-UHFFFAOYSA-N
Synonym oxamic hydrazide, semioxamazide, semioxamazid, aminooxamide, oxamic acid hydrazide, oxamichydrazide, n-aminooxamide, 1-hydrazinecarbonyl formamide, oxamic acid, hydrazide, unii-bet2ubd6nv
PubChem CID 68200
IUPAC Name 2-hydrazinyl-2-oxoacetamide
SMILES C(=O)(C(=O)NN)N

Specifications

Color White
Quantity 5 g
UN Number 1325
Formula Weight 103.08
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Oxamic Hydrazide
RTECSNumber AF2620000
TSCA No