missing translation for 'onlineSavingsMsg'
Learn More

Oxamic acid sodium salt, 98%

Catalog No. AAA1653206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1653206 5 g
AAA1653214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1653206 Supplier Thermo Scientific Chemicals Supplier No. A1653206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 565-73-1 | C2H2NNaO3 | 111.032 g/mol

Sodium oxamate, an inhibitor of LDH-C4, prevented the AR induced by lysophosphatidylcholine (LPC) or NADH. Oxamic acid sodium salt is an lactic acid dehydrogenase. Sperm suspension and cytosol fraction activities were inhibited by sodium oxamate. As cancer cells are often dependent on glycolysis for ATP production, sodium oxamate has implications as an anticancer compound.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium oxamate, an inhibitor of LDH-C4, prevented the AR induced by lysophosphatidylcholine (LPC) or NADH. Oxamic acid sodium salt is an lactic acid dehydrogenase. Sperm suspension and cytosol fraction activities were inhibited by sodium oxamate. As cancer cells are often dependent on glycolysis for ATP production, sodium oxamate has implications as an anticancer compound.

Solubility
Soluble in water (10 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 565-73-1
Molecular Formula C2H2NNaO3
Molecular Weight (g/mol) 111.032
MDL Number MFCD00044553
InChI Key RQVZIJIQDCGIKI-UHFFFAOYSA-M
Synonym sodium oxamate, oxamic acid sodium salt, sodium 2-amino-2-oxoacetate, oxamic acid, sodium salt, sodium amino oxo acetate, acetic acid, aminooxo-, monosodium salt, oxalic acid monoamide sodium salt, acetic acid, 2-amino-2-oxo-, sodium salt 1:1, oxamic acid sodium, lopac-o-2751
PubChem CID 5242
IUPAC Name sodium;oxamate
SMILES C(=O)(C(=O)[O-])N.[Na+]

Specifications

Melting Point >300°C
Quantity 5 g
Beilstein 6538153
Solubility Information Soluble in water (10mg/ml).
Formula Weight 111.03
Percent Purity 98%
Chemical Name or Material Oxamic acid sodium salt
EINECSNumber 209-290-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only