missing translation for 'onlineSavingsMsg'
Learn More

Oxalyl Chloride 98.0+%, TCI America™

Catalog No. O008225G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O008225G 25 g
O0082100G 100 g
O0082500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. O008225G Supplier TCI America Supplier No. O008225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 79-37-8
Molecular Formula C2Cl2O2
Molecular Weight (g/mol) 126.92
MDL Number MFCD00000704
InChI Key CTSLXHKWHWQRSH-UHFFFAOYSA-N
Synonym oxalyl chloride, ethanedioyl dichloride, oxalic dichloride, oxaloyl chloride, oxalic acid dichloride, oxalic acid chloride, ethanedioyl chloride, oxalylchloride, unii-r4y96317dw, cocl 2
PubChem CID 65578
IUPAC Name oxalyl dichloride
SMILES C(=O)(C(=O)Cl)Cl

Specifications

Melting Point -12°C
Color Yellow
Boiling Point 64°C
Quantity 25 g
UN Number 2922
Formula Weight 126.92
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Oxalyl Chloride
EINECSNumber (9)-2022
RTECSNumber KI2950000
TSCA Yes