missing translation for 'onlineSavingsMsg'
Learn More

Oxalyl chloride, 98%

Catalog No. p-177712
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC129610250 25 g Glass bottle
AC129611000 100 g Glass bottle
AC129610010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC129610250 Supplier Thermo Scientific Chemicals Supplier No. 129610250

CAS: 79-37-8 | C2Cl2O2 | 126.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent, Oxidation reagent

Chemical Identifiers

CAS 79-37-8 , 75-44-5 , 76-02-8
Molecular Formula C2Cl2O2
Molecular Weight (g/mol) 126.93
MDL Number MFCD00000704
InChI Key CTSLXHKWHWQRSH-UHFFFAOYSA-N
Synonym oxalyl chloride, ethanedioyl dichloride, oxalic dichloride, oxaloyl chloride, oxalic acid dichloride, oxalic acid chloride, ethanedioyl chloride, oxalylchloride, unii-r4y96317dw, cocl 2
PubChem CID 65578
IUPAC Name oxalyl dichloride
SMILES C(=O)(C(=O)Cl)Cl

Specifications

Melting Point -12.0°C
Color Colorless to Yellow
Density 1.4780g/mL
Boiling Point 63.0°C to 64.0°C (763.0 mmHg)
Flash Point >100°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4285 to 1.4305
Linear Formula ClCOCOCl
Quantity 25 g
Beilstein 02, 542
Fieser 01,767; 02,301; 03,216; 04,361; 05,481; 06,424; 07,257; 08,366; 11,379; 14,150; 16,149; 17,241
Specific Gravity 1.478
Merck Index 15, 7013
Solubility Information Solubility in water: reacts. Other solubilities: soluble in the main common organic solvents :, methylene chloride; dichloroethane; chlorobenzene;, ethylacetate; hexane; acetone; toluene; ether;, benzene; chloroform and ether
Formula Weight 126.93
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Oxalyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if inhaled.
Toxic if swallowed.
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts violently with water.
Contact with w
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Handle under inert gas.
Protect from moisture.
Brush off loose particles from skin.
Immerse in cool water/wrap in wet bandages.
IF INHA
EINECSNumber 201-200-2
RTECSNumber KI2950000
TSCA TSCA

RUO – Research Use Only