missing translation for 'onlineSavingsMsg'
Learn More

Oxalic Acid Dihydrate, TraceSELECT™, ≥99.9999% (Metals basis), Solstice

Catalog No. 60047753
Change view
Click to view available options
Quantity:
100 g
Packaging:
Poly bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60047753 100 g Poly bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60047753 Supplier Solstice Supplier No. 93722100G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

TraceSELECT™,≥99.9999% (metals basis)

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions

Chemical Identifiers

CAS 6153-56-6
Molecular Formula C2H6O6
Molecular Weight (g/mol) 126.064
MDL Number MFCD00149102
InChI Key GEVPUGOOGXGPIO-UHFFFAOYSA-N
Synonym oxalic acid dihydrate, ethanedioic acid, dihydrate, unii-0k2l2ij59o, ethanedioic acid dihydrate, oxalic acid, dihydrate, oxalicacid dihydrate, dihydrate oxalic acid, oxalic acid di-hydrate, pubchem17416, acmc-209mtm
PubChem CID 61373
IUPAC Name oxalic acid;dihydrate
SMILES C(=O)(C(=O)O)O.O.O

Specifications

pH 1 (126.1g/L at 25°C)
Melting Point 104°C to 106°C
Color Colorless
Quantity 100 g
Packaging Poly bottle
Physical Form Crystalline
Linear Formula HO2CCO2H · 2H2O
Vapor Density 4.4 (vs air)
UN Number NONH for all modes of transport
Vapor Pressure 0.01mmHg (20°C)
Beilstein 3679436
Formula Weight 126.07g/mol
Percent Purity ≥99.9999% (metals basis)
Grade TraceSELECT
Chemical Name or Material Oxalic acid dihydrate
Show More Show Less
Hazard Statement P280-P305 + P351 + P338
RTECSNumber 205-634-3