missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxalic Acid Dihydrate, MP Biomedicals™
$295.14 - $295.14
Chemical Identifiers
| CAS | 6153-56-6 |
|---|---|
| Molecular Formula | C2H6O6 |
| InChI Key | GEVPUGOOGXGPIO-UHFFFAOYSA-N |
| Synonym | oxalic acid dihydrate, ethanedioic acid, dihydrate, unii-0k2l2ij59o, ethanedioic acid dihydrate, oxalic acid, dihydrate, oxalicacid dihydrate, dihydrate oxalic acid, oxalic acid di-hydrate, pubchem17416, acmc-209mtm |
| PubChem CID | 61373 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN15255290
|
MP Biomedicals Inc
0215255290 |
500 g |
N/A
|
|
|||||
Chemical Identifiers
| 6153-56-6 | |
| GEVPUGOOGXGPIO-UHFFFAOYSA-N | |
| 61373 |
| C2H6O6 | |
| oxalic acid dihydrate, ethanedioic acid, dihydrate, unii-0k2l2ij59o, ethanedioic acid dihydrate, oxalic acid, dihydrate, oxalicacid dihydrate, dihydrate oxalic acid, oxalic acid di-hydrate, pubchem17416, acmc-209mtm |
Specifications
| 6153-56-6 | |
| C2H6O6 | |
| GEVPUGOOGXGPIO-UHFFFAOYSA-N | |
| oxalic acid;dihydrate | |
| 61373 | |
| 500 g | |
| Oxalic acid 2-hydrate, For ACS analysis |
| White | |
| oxalic acid dihydrate, ethanedioic acid, dihydrate, unii-0k2l2ij59o, ethanedioic acid dihydrate, oxalic acid, dihydrate, oxalicacid dihydrate, dihydrate oxalic acid, oxalic acid di-hydrate, pubchem17416, acmc-209mtm | |
| C(=O)(C(=O)O)O.O.O | |
| 126.064 | |
| 126.07 | |
| Crystalline |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.