missing translation for 'onlineSavingsMsg'
Learn More

Oxalic Acid, Dihydrate, Crystal, ACS, 99.5-102.5%, Spectrum™ Chemical

Catalog No. p-7165983
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
3 kg
12 kg
25 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612104 125 g Poly Bottle
18612105 500 g Poly Bottle
18612106 2.5 kg Poly Bottle
18612107 3 kg Poly Bottle
18612108 12 kg Poly Pail
18612109 25 kg Fiber Drum
18612110 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18612104 Supplier Spectrum Chemical Mfg Cor Supplier No. O1025125GM
Only null left

Chemical Identifiers

CAS 6153-56-6
Molecular Formula C2H6O6
Molecular Weight (g/mol) 126.06
InChI Key GEVPUGOOGXGPIO-UHFFFAOYSA-N
IUPAC Name oxalic acid dihydrate
SMILES O.O.OC(=O)C(O)=O

Specifications

CAS Max % 1
Percent Purity 99.5 to 102.5%
Grade ACS
Quantity 125 g
Packaging Poly Bottle
Residue after Ignition 0.0001

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.