missing translation for 'onlineSavingsMsg'
Learn More

Oxalic Acid Dihydrate, ACS Reagent, Reagents

Oxalic Acid Dihydrate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Supplier:  Labchem Inc C111050025F4

 View more versions of this product

Catalog No. 01-338-167

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Oxalic Acid Dihydrate, ACS Reagent
6153-56-6
C2H6O6
GEVPUGOOGXGPIO-UHFFFAOYSA-N
oxalic acid dihydrate
ACS
Fiber Drum
1.653
101°C to 102°C
Colorless/White
Ethanedioic Acid dihydrate
O.O.OC(=O)C(O)=O
126.06
25 kg
Solid

Chemical Identifiers

6153-56-6
126.06
Ethanedioic Acid dihydrate
O.O.OC(=O)C(O)=O
C2H6O6
GEVPUGOOGXGPIO-UHFFFAOYSA-N
oxalic acid dihydrate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions