missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxalic Acid Dihydrate, ACS Reagent, Reagents
Oxalic Acid Dihydrate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C1110500500A5
Specifications
| Oxalic Acid Dihydrate, ACS Reagent | |
| 6153-56-6 | |
| C2H6O6 | |
| GEVPUGOOGXGPIO-UHFFFAOYSA-N | |
| oxalic acid dihydrate | |
| ACS | |
| Poly Bottle | |
| 1.653 |
| 101°C to 102°C | |
| Colorless/White | |
| Ethanedioic Acid dihydrate | |
| O.O.OC(=O)C(O)=O | |
| 126.06 | |
| 500 g | |
| Solid |
Chemical Identifiers
| 6153-56-6 | |
| 126.06 | |
| Ethanedioic Acid dihydrate | |
| O.O.OC(=O)C(O)=O |
| C2H6O6 | |
| GEVPUGOOGXGPIO-UHFFFAOYSA-N | |
| oxalic acid dihydrate |