missing translation for 'onlineSavingsMsg'
Learn More

Oxalic Acid Dihydrate, ACS Reagent, Reagents

Catalog No. 01338168
Change view
Click to view available options
Quantity:
500 g
12 kg
25 kg
50 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01338168 12 kg
01338167 25 kg
01338166 500 g
01338165 50 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01338168 Supplier Labchem Inc Supplier No. C111050012A5

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Oxalic Acid Dihydrate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 6153-56-6
Molecular Formula C2H6O6
Molecular Weight (g/mol) 126.06
InChI Key GEVPUGOOGXGPIO-UHFFFAOYSA-N
Synonym Ethanedioic Acid dihydrate
IUPAC Name oxalic acid dihydrate
SMILES O.O.OC(=O)C(O)=O

Specifications

Melting Point 101°C to 102°C
Color Colorless/White
Quantity 12 kg
Packaging Poly Pail
Physical Form Solid
Specific Gravity 1.653
Grade ACS
Chemical Name or Material Oxalic Acid Dihydrate, ACS Reagent