missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxalic Acid Dihydrate, ACS Reagent Grade, Ricca Chemical
$290.94 - $290.94
Chemical Identifiers
| CAS | 6153-56-6 |
|---|---|
| Molecular Formula | C2H6O6 |
| InChI Key | GEVPUGOOGXGPIO-UHFFFAOYSA-N |
| Synonym | oxalic acid dihydrate, ethanedioic acid, dihydrate, unii-0k2l2ij59o, ethanedioic acid dihydrate, oxalic acid, dihydrate, oxalicacid dihydrate, dihydrate oxalic acid, oxalic acid di-hydrate, pubchem17416, acmc-209mtm |
| PubChem CID | 61373 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
RDCO0200500
|
Ricca Chemical Company
RDCO0200500B1 |
500 g | Poly Wide Mouth Bottle |
Each for $290.94
|
|
||||
Chemical Identifiers
| 6153-56-6 | |
| GEVPUGOOGXGPIO-UHFFFAOYSA-N | |
| 61373 |
| C2H6O6 | |
| oxalic acid dihydrate, ethanedioic acid, dihydrate, unii-0k2l2ij59o, ethanedioic acid dihydrate, oxalic acid, dihydrate, oxalicacid dihydrate, dihydrate oxalic acid, oxalic acid di-hydrate, pubchem17416, acmc-209mtm |
Specifications
| 6153-56-6 | |
| 100.0 | |
| oxalic acid dihydrate, ethanedioic acid, dihydrate, unii-0k2l2ij59o, ethanedioic acid dihydrate, oxalic acid, dihydrate, oxalicacid dihydrate, dihydrate oxalic acid, oxalic acid di-hydrate, pubchem17416, acmc-209mtm | |
| C(=O)(C(=O)O)O.O.O | |
| 126.064 | |
| 99.5 to 102.5% | |
| 500 g | |
| Oxalic Acid, Dihydrate |
| 98.0 | |
| C2H6O6 | |
| GEVPUGOOGXGPIO-UHFFFAOYSA-N | |
| oxalic acid;dihydrate | |
| 61373 | |
| ACS | |
| Poly Wide Mouth Bottle |
Safety and Handling
ShelfLife : 60 months