Learn More
Oxalic acid dihydrate, ACS, 99.5-102.5%
CAS: 6153-56-6 | C2H6O6 | 126.064 g/mol
Supplier: Thermo Scientific Chemicals 033262A1
Description
As an analytical reagent, in dye and paint industries, in polymers, in metal treatment, in cleaning and textile finishing and as a reagent for dehydration and condensation Oxalic acid dihydrate acts as an analytical reagent, as a reducing agent for photographic films and as mordant in the dyeing process. It is used as a plating agent, a surface treating agent, a grinding agent, a precipitating agent in rare-earth mineral processing, a bleaching agent in textile and a purifying agent in the pharmaceutical industry. It plays an important role in antibiotic medication like oxytetracycline and chloramphenicol. It finds application in water treatment plants to remove calcium waste water. It is also used as a buffer in chromatographic separation as well as bleach for wood, removing black stains caused by water penetration.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- As an analytical reagent, in dye and paint industries, in polymers, in metal treatment, in cleaning and textile finishing, and as a reagent for dehydration and condensation
Specifications
| Oxalic acid dihydrate | |
| 101°C to 105°C | |
| White | |
| C2H6O6 | |
| MFCD00149102 | |
| 3679436 | |
| oxalic acid dihydrate, ethanedioic acid, dihydrate, unii-0k2l2ij59o, ethanedioic acid dihydrate, oxalic acid, dihydrate, oxalicacid dihydrate, dihydrate oxalic acid, oxalic acid di-hydrate, pubchem17416, acmc-209mtm | |
| GEVPUGOOGXGPIO-UHFFFAOYSA-N | |
| oxalic acid;dihydrate | |
| 61373 | |
| 99.5 to 102.5% | |
| 1.65 g/mL | |
| Odorless |
| 1.3 | |
| 6153-56-6 | |
| 99.5-102.5% | |
| HO2CCO2H·2H2O | |
| UN3261 | |
| 14,6911 | |
| Soluble in water,ethanol,ether and glycerol; Insoluble in chloroform. | |
| C(=O)(C(=O)O)O.O.O | |
| 126.064 | |
| 126.07 (90.04 Anhydrous) | |
| ACS | |
| 1 kg | |
| Crystalline |
Chemical Identifiers
| 6153-56-6 | |
| 126.064 | |
| GEVPUGOOGXGPIO-UHFFFAOYSA-N | |
| 61373 | |
| C(=O)(C(=O)O)O.O.O |
| C2H6O6 | |
| MFCD00149102 | |
| oxalic acid dihydrate, ethanedioic acid, dihydrate, unii-0k2l2ij59o, ethanedioic acid dihydrate, oxalic acid, dihydrate, oxalicacid dihydrate, dihydrate oxalic acid, oxalic acid di-hydrate, pubchem17416, acmc-209mtm | |
| oxalic acid;dihydrate |
Safety and Handling
GHS H Statement
H318-H302-H312
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
P260-P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P310-P312-P314-P330-P363-P501c
H302+H312-H318-H373
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 205-634-3
RTECSNumber : KI1600000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only