missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Oxalic acid

Numéro de catalogue. 1941311KG
Encompass
Modifier l'affichage
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
1941311KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 1941311KG Fournisseur Sigma Aldrich Code fournisseur 1941311KG
Il en reste null

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Oxalic acid

Spécifications

Melting Point 189.5°C (lit.)
CAS 144-62-7
Molecular Formula C2H2O4
Linear Formula HO2CCO2H
MDL Number MFCD00002573
Molecular Weight (g/mol) 90.03
Percent Purity 98%
Density 1.65 g/cm3 (at 18.5 C)
Quantity 1 kg