missing translation for 'onlineSavingsMsg'
Learn More

Oxalacetic Acid 97.0+%, TCI America™

Catalog No. p-7134171
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O00725G 5 g
O007225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O00725G Supplier TCI America Supplier No. O00725G
Only null left

Chemical Identifiers

CAS 328-42-7
Molecular Formula C4H4O5
Molecular Weight (g/mol) 132.071
MDL Number MFCD00002592
InChI Key KHPXUQMNIQBQEV-UHFFFAOYSA-N
Synonym oxalacetic acid, oxaloacetic acid, 2-oxosuccinic acid, oxaloacetate, ketosuccinic acid, oxosuccinic acid, 2-ketosuccinic acid, butanedioic acid, oxo, oxobutanedioic acid, oxalacetate
PubChem CID 970
ChEBI CHEBI:30744
IUPAC Name 2-oxobutanedioic acid
SMILES C(C(=O)C(=O)O)C(=O)O

Specifications

Color White-Yellow
Quantity 5 g
UN Number 3261
Formula Weight 132.07
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Oxalacetic Acid
TSCA Yes
Recommended Storage Refrigerator