missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxacillin Sodium Salt Monohydrate 98.0+%, TCI America™
$211.77 - $621.84
Chemical Identifiers
| CAS | 7240-38-2 |
|---|---|
| Molecular Formula | C19H23N3NaO6S |
| Molecular Weight (g/mol) | 444.458 |
| MDL Number | MFCD00167146 |
| InChI Key | VKNIYYBMVNGVMD-MBGFYOAHSA-N |
| PubChem CID | 131673944 |
| IUPAC Name | (2S,5R,6R)-3,3-dimethyl-6-[(5-methyl-3-phenyl-1,2-oxazole-4-carbonyl)amino]-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;sodium;hydrate |
| SMILES | [HH].CC1=C(C(=NO1)C2=CC=CC=C2)C(=O)NC3C4N(C3=O)C(C(S4)(C)C)C(=O)O.O.[Na] |
Chemical Identifiers
| 7240-38-2 | |
| 444.458 | |
| VKNIYYBMVNGVMD-MBGFYOAHSA-N | |
| (2S,5R,6R)-3,3-dimethyl-6-[(5-methyl-3-phenyl-1,2-oxazole-4-carbonyl)amino]-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;sodium;hydrate |
| C19H23N3NaO6S | |
| MFCD00167146 | |
| 131673944 | |
| [HH].CC1=C(C(=NO1)C2=CC=CC=C2)C(=O)NC3C4N(C3=O)C(C(S4)(C)C)C(=O)O.O.[Na] |
Specifications
| 7240-38-2 | |
| White-Yellow | |
| MFCD00167146 | |
| VKNIYYBMVNGVMD-MBGFYOAHSA-N | |
| (2S,5R,6R)-3,3-dimethyl-6-[(5-methyl-3-phenyl-1,2-oxazole-4-carbonyl)amino]-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;sodium;hydrate | |
| 131673944 | |
| ≥98.0% (HPLC) | |
| Oxacillin Sodium Salt Monohydrate |
| 188°C | |
| C19H23N3NaO6S | |
| 5 g | |
| [HH].CC1=C(C(=NO1)C2=CC=CC=C2)C(=O)NC3C4N(C3=O)C(C(S4)(C)C)C(=O)O.O.[Na] | |
| 444.458 | |
| 423.42 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator