missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Owl™ VEP-3 Large Tank Electroblotting System

Catalog No. 09528141
Change view
Click to view available options
Type:
Electroblotting System
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
09528141 Electroblotting System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09528141 Supplier Thermo Scientific™ Supplier No. VEP 3

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rapidly transfer high molecular weight proteins with the Thermo Scientific™ Owl™ VEP-3 Large Tank Electroblotting System.

Features:
  • Removable cooling block permits efficient heat transfer.
  • Coded blotting cassettes ensure proper anode/cathode orientation; load easily into buffer chamber.
  • Stir bars (sold separately) may be added to bottom of buffer chamber to circulate buffer.
  • Cassettes allow easy loading of the gel sandwich and locks securely for loading into the buffer chamber.

Includes:

  • Buffer chamber with stainless-steel cathode plate and titanium anode plate
  • Safety interlocking lid with attached power supply leads and cooling module
  • Two blotting cassettes with foam pads
  • 25 sheets of 20W × 18.5cmL (7.88 × 7.25 in.) blotting filter paper

Specifications

Gel Size 20 x 18.5 cm
Product Line Owl
Type Electroblotting System
Volume (English) Buffer 1.05 gal.
Volume (Metric) Buffer 4 L
Dimensions (L x W x H) 3.5 x 11.8 x 30 in. (9 x 30 x 30 cm)
Length (English) 3.5 in.
Length (English) Gel 7.8 in.
Length (Metric) 9 cm
Length (Metric) Gel 20 cm
Width (English) 11.8 in.
Width (English) Gel 7.2 in.
Width (Metric) 30 cm
Width (Metric) Gel 18.5 cm
Height (English) 30 in.
Height (Metric) 30 cm
Show More Show Less