missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Owl™ VEP-2 Mini Tank Electroblotting System

Catalog No. 09528135
Change view
Click to view available options
Type:
Electroblotting System
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
09528135 Electroblotting System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09528135 Supplier Thermo Scientific™ Supplier No. VEP 2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Produce uniform, reproducible protein transfers for up to four polyacrylamide minigels with the Thermo Scientific™ Owl™ VEP-2 Mini Tank Electroblotting System.

Produce uniform, reproducible protein transfers for up to four polyacrylamide minigels with the Thermo Scientific™ Owl™ VEP-2 Mini Tank Electroblotting System, ideal for fast, efficient transfer of proteins over a wide molecular weight range.

Features:
  • Tank-style electroblotting system
  • Integrated cooling base enables temperature-controlled runs
  • Stir bars (not included) may be added to bottom of buffer chamber for increased buffer circulation

Includes:

  • Buffer chamber with platinum wire electrode panels and cooling base
  • Safety interlocking lid with attached power supply leads
  • Four blotting cassettes with foam pads and cassette clips
  • Six sheets of blotting filter paper

Specifications

Gel Size Mini (9 in. x 9 in.)
Product Line Owl
Type Electroblotting System
Volume (English) Buffer 0.3 gal.
Volume (Metric) Buffer 1.4 L
Dimensions (L x W x H) 7.06 x 5.13 x 6.5 in. (18 x 13 x 16.5 cm)
Length (English) 7.06 in.
Length (English) Gel 3.5 in.
Length (Metric) 18 cm
Length (Metric) Gel 9 cm
Width (English) 5.13 in.
Width (English) Gel 3.5 in.
Width (Metric) 13 cm
Width (Metric) Gel 9 cm
Height (English) 6.5 in.
Height (Metric) 16.5 cm
Show More Show Less