missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Owl™ HEP Series Semidry Electroblotting Systems

Catalog No. OWHEP1
Change view
Click to view available options
Dimensions (L x W x H):
25 x 25 x 5 cm
Capacity:
Transfer area: 20 x 20 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H) Capacity
OWHEP1 25 x 25 x 5 cm Transfer area: 20 x 20 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OWHEP1 Supplier Thermo Scientific™ Supplier No. HEP 1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Provide faster blots with higher efficiency using Thermo Scientific™ Owl™ HEP Series Semidry Electroblotting Systems.

Provide faster blots with higher efficiency using Thermo Scientific™ Owl™ HEP Series Semidry Electroblotting Systems. Transfers can be done in as little as 15 minutes.

  • Fast, efficient transfer of proteins and nucleic acids to membranes
  • Buffer contained within blotting paper is sufficient for efficient transfer of molecules
  • Base contains stainless-steel cathode plate; lid has platinum/titanium anode plate
  • Ideal for proteins less than 150 kDa

Includes:

  • Base and lid
  • Power supply leads
  • Heavy-duty knobs

Specifications

Capacity Transfer area: 20 x 20 cm
Product Line Owl
Type Electroblotting System
Dimensions (L x W x H) 25 x 25 x 5 cm