Learn More
Thermo Scientific™ Owl™ Gel Staining Box
Catalog No. OWGSB3
Change view
Click to view available options
Quantity:
Pk. of 5 boxes
Catalog No. Quantity
OWGSB3 Pk. of 5 boxes
1 options
Catalog No. OWGSB3 Supplier Thermo Scientific™ Supplier No. GSB3
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Stain, fix and de-stain in the same container with Thermo Scientific™ Owl™ Gel Staining Boxes, offering a convenient, conventional staining method.
Stain, fix and de-stain in the same container with Thermo Scientific™ Owl™ Gel Staining Boxes, offering a convenient, conventional staining method. Boxes hold up to 125mL of liquid and one gel when used on a shaking platform.
  • Trays available in convenient size to hold the proper volume of staining reagents
  • Grooved lid enables proper orientation and stability when stacking
  • Made of clear plastic, so gels can be viewed directly in the tray
Ordering Information:
Package of five
Specifications
Description Gel Staining Box; Pack of 5
Quantity Pk. of 5 boxes
Volume (Metric) 125 mL
Length (Metric) 11 cm
Width (Metric) 11 cm
Height (Metric) 3 cm