missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Owl™ EasyCast™ B1A Mini Gel Electrophoresis Systems

Catalog No. OWB1ABP
Change view
Click to view available options
Includes:
EasyCast w/ Buffer Exchange Ports
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
OWB1ABP EasyCast w/ Buffer Exchange Ports
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OWB1ABP Supplier Thermo Scientific™ Supplier No. B1A BP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cast and run a gel in the buffer chamber and achieve flat, even banding patterns on a consistent basis with these B1A Mini Gel Electrophoresis systems.

Cast and run a gel in the buffer chamber and achieve flat, even banding patterns on a consistent basis with Thermo Scientific™ Owl™ EasyCast™ B1A Mini Gel Electrophoresis Systems. The ideal size for every lab, this system determines molecular weight ranges of PCR fragments or larger sized DNA molecules.

Features:
  • Two comb slots on gel tray double sample capacity by doubling number of sample wells.
  • UVT gel trays are silk screened with a florescent ruler for easy measurement of bands.
  • Leakproof casting allows a gel to be cast and run in the same chamber with no tape or additional parts required
  • Run from five to 24 samples in 15 minutes.

Ordering Information:

Available with optional buffer exchange ports (B1A-BP) for use with an external recirculation pump.

Includes:

  • Main buffer chamber
  • Thermo Scientific™ SuperSafe™ lid with attached power supply leads
  • Thermo Scientific™ EasyCast gasketed UVT gel tray
  • Two heavy-duty, one-piece, double-sided combs: 6 & 10 well, 1.0/1.5mm thick

Specifications

Certifications/Compliance CE marked
Gel Size Mini
Product Line Owl
Type Mini Gel Electrophoresis System
Includes EasyCast w/ Buffer Exchange Ports
Sample Size 5 to 24
Volume (Metric) Buffer 400 mL
Dimensions (L x W x H) 6.31 x 4.13 x 3.75 in. (16 x 10.5 x 9.5 cm)
Length (English) 6.31 in.
Length (Metric) 16 cm
Length (Metric) Gel 8 cm
Width (English) 4.13 in.
Width (Metric) 10.5 cm
Width (Metric) Gel 7 cm
Height (English) 3.75 in.
Height (Metric) 9.5 cm
Show More Show Less