missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Owl™ EasyCast™ B1 Mini Gel Electrophoresis Systems

Catalog No. OWB1BP
Change view
Click to view available options
Includes:
Buffer Exchange Ports
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
OWB1BP Buffer Exchange Ports
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OWB1BP Supplier Thermo Scientific™ Supplier No. B1 BP
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cast and run a gel in the same chamber with no tape or additional parts required with the leakproof casting of mini gel electrophoresis systems.

Cast and run a gel in the same chamber with no tape or additional parts required with the leakproof casting of Thermo Scientific™ Owl™ EasyCast™ B1 Mini Gel Electrophoresis Systems. Durability and ease of use makes this the perfect choice for students and high volume researchers.

Features:

  • System offers a wide range of combs.
  • Two comb slots on gel tray double sample capacity by doubling number of sample wells.
  • UVT gel trays are silk screened with a florescent ruler for easy measurement of bands.

Includes:

  • Main buffer chamber
  • Thermo Scientific™ SuperSafe™ lid with attached power supply leads
  • Thermo Scientific™ EasyCast™ Gasketed UVT gel tray
  • Two Combs: 10 & 14 Well, Double-sided, 1.0/1.5mm thick

Ordering Information:

Available with optional buffer exchange ports, (B1-BP), for use with an external recirculation pump.

Warranty and Service Offering:

  • Three years

Specifications

Certifications/Compliance CE marked
Gel Size Mini
Product Line Owl
Type Mini Gel Electrophoresis System
Includes Buffer Exchange Ports
Sample Size 5 to 34
Volume (Metric) Buffer 600 mL
Dimensions (L x W x H) 8.63 x 5.88 x 3.75 in. (22 x 15 x 9.5 cm)
Length (English) 8.63 in.
Length (Metric) 22 cm
Length (Metric) Gel 11 cm
Width (English) 5.88 in.
Width (Metric) 15 cm
Width (Metric) Gel 9 cm
Height (English) 3.75 in.
Height (Metric) 9.5 cm
Show More Show Less