missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Owl™ D2 Wide-Gel Electrophoresis System

Catalog No. OWD2BP
Change view
Click to view available options
Type:
Wide w/Buffer Exchange Ports
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
OWD2BP Wide w/Buffer Exchange Ports
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OWD2BP Supplier Thermo Scientific™ Supplier No. D2BP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Run 10 to 80 samples on one gel with this wide-gel electrophoresis system.

Run 10 to 80 samples on one gel with the Thermo Scientific™ Owl™ D2 Wide-Gel Electrophoresis System.

  • Wide gel size allows 15 or 30 samples to be loaded per row with a micro well comb.
  • Using special combs and a multichannel pipetter, samples may be loaded directly from a 96-well plate, eight or 12 at a time.
  • System is excellent for screening PCR samples or routine DNA fragment analysis.
  • Small footprint saves space on lab bench.
  • Incorporates Thermo Scientific™ EasyCast™ casting design with external caster—gel tray is fitted with gasket, allowing it to fit snugly into caster and providing a leakproof seal without tape.
  • Electrode placement allows gel to be run quickly without compromising resolution.

Includes:

  • Buffer chamber
  • Thermo Scientific™ SuperSafe™ lid with attached power supply leads
  • EasyCast gasketed UVT gel tray
  • External multiple gel caster
  • Four combs (two 30-well and two 40-well, 1.5mm thick)

Specifications

Certifications/Compliance CE marked
Description Complete System with Buffer Exchange Ports
Product Line Owl
Type Wide w/Buffer Exchange Ports
Sample Size 10 to 80
Volume (Metric) Buffer 600 mL
Dimensions (L x W x H) 7.38 x 7.75 x 4 in. (19 x 20 x 10 cm)
Length (English) 7.38 in.
Length (Metric) 19 cm
Length (Metric) Gel 14.4 cm
Width (English) 7.75 in.
Width (English) Gel 6.5
Width (Metric) 20 cm
Width (Metric) Gel 10.2 cm
Height (English) 4 in.
Height (Metric) 10 cm
Max. Voltage 150 V
Show More Show Less