missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Owl™ Aluminum-Backed Sequencers
Description
Easily set up applications with even heat distribution for smile-free results with manual Thermo Scientific™ Owl™ Aluminum-Backed Sequencers.
Features:
- Floating aluminum heat sink reduces chance of plates cracking during runs.
- Upper buffer chamber (UBC) requires minimal buffer and is easily drained via inline drain port into removable bottle.
- Lower buffer chamber (LBC) is removable for simple buffer disposal.
- Side clamps press gel plates against gasket seated on UBC, applying even pressure across plate and forming a leak-proof seal resulting in flat, even banding patterns.
- Clamps slide over glass plates for easy setup and removal of gels.
Includes:
- Removable LBC
- Heat sink
- Two side clamps
- Two blank glass plates
- Two notched glass plates
- Two sharktooth combs (78 teeth, 0.4mm thick)
- Well comb (40 well, 0.4mm thick)
- Two spacer sets (0.4mm)
- Adhesive temperature indicator
- UBC with removable drain bottle (with safety interlocking lid and attached power supply lead)
Specifications
Specifications
| Dimensions (L x W x H) Exterior | 10.5 x 9 x 19.5 in. (27 x 23 x 50cm) |
| Format | 40-well |
| Height (English) Exterior | 19.5 in. |
| Height (Metric) Exterior | 50 cm |
| Length (English) Exterior | 10.5 in. |
| Length (English) Gel | 17.716 in. |
| Length (Metric) Exterior | 27 cm |
| Length (Metric) Gel | 45 cm |
| Volume (English) Buffer | 27 fl oz. |
| Volume (Metric) Buffer | 800mL |
| Show More |