missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Owl™ A3-1 Large-Gel Electrophoresis System

Catalog No. OWA31
Change view
Click to view available options
Gel Size:
23 x 40 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Gel Size
OWA31 23 x 40 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OWA31 Supplier Thermo Scientific™ Supplier No. A3 1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Conduct plant genomics studies with Thermo Scientific™ Owl™ A3-1 Large-Gel Electrophoresis System.

Features:
  • Horizontal system can run from 25 to 600 samples on one gel
  • Gel can be cast in varying lengths to help conserve agarose when fewer samples need to be run
  • Built-in buffer exchange ports allow for buffer recirculation on extended runs
  • UVT gel tray has gasketed end gates for leakproof seal without tape
  • Built-in leveling ensures flat, uniformly thick gels
  • Large number of comb slots provides sample number and resolving distance flexibility
  • Microwell-format combs provide fast, error-free sample loading with a multichannel pipetter
  • Samples may be loaded directly from a 96-well plate, 8 or 12 at a time

Ordrring Information:

Customized combs are available (Order SKU # XCM). Please contact your Customer Service or Sales Representative for further details.

Includes:

Buffer chamber with three-point leveling base, SuperSafe™ lid with attached power supply leads, UVT gel tray, four combs (two 25- and two 50-well), built-in buffer exchange ports and bubble level

Specifications

Certifications/Compliance CE marked
Description Gel size: 23 x 40cm; Buffer volume: 4.5L; L x W x H: 53 x 32 x 11cm
Gel Size 23 x 40 cm
Product Line Owl
Type Large Gel Electrophoresis System
Sample Size 25 to 600
Volume (Metric) Buffer 4.5 L
Dimensions (L x W x H) 20.63 x 12.5 x 4.25 in. (53 x 32 x 11 cm)
Length (English) 20.63 in.
Length (Metric) 53 cm
Length (Metric) Gel 40 cm
Width (English) 12.5 in.
Width (Metric) 32 cm
Width (Metric) Gel 23 cm
Height (English) 4.25 in.
Height (Metric) 11 cm
Max. Voltage 250 V
Show More Show Less