missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Owen Mumford™ Unistik™ 3 Single-Use Safety Lancets
Description
- Requires no cocking procedure – just twist off protective sterile cap and use
- Convex platform requires less technique and training for use
- Ergonomically designed with special indentions on each side for finger placement
- Needle point is hidden before use and automatically retracts after use to avoid accidental needlestick injuries and cross-infection
- Rendered inoperable after use, providing highest level of safety for healthcare workers and patients and meeting OSHA standards
- Visual lockout feature clearly indicates that device has been used and is ready for disposal in a sharps container
- High-flow volume suitable for cholesterol, hemoglobin and cardiac enzyme testing
US Patents 5,487,748; 6,258,112; 6,764,496
Specifications
Specifications
| Disposable | Single-use |
| Depth (English) | 0.07 in. |
| Needle Gauge | 21g |
| Product Type | Unistik™ 3 Safety Lancet |
| Quantity | 1000 Case |
| Depth (Metric) | 2mm |
| Color | Orange |
| For Use With (Application) | For ward blood glucose monitoring, HbA1c |