missing translation for 'onlineSavingsMsg'
Learn More

Owen Mumford™ Unistik™ 3 Single-Use Safety Lancets

Catalog No. 2202932
Change view
Click to view available options
Color:
Lavendar
Orange
Depth (Metric):
1.8mm
2mm
Needle Gauge:
21g
28g
Quantity:
100 Pack
1000 Case
200 Pack
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Depth (Metric) Needle Gauge Quantity
2202932 Lavendar 1.8mm 28g 1000 Case
22023380 Orange 2mm 21g 1000 Case
22023381 Orange 2mm 21g 100 Pack
22023382 Orange 2mm 21g 200 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 2202932 Supplier Owen Mumford™ Supplier No. AT 1043

Unique design features a side release button and pre-set lancet speed to ensure maximum comfort for patient and healthcare worker

  • Requires no cocking procedure – just twist off protective sterile cap and use
  • Convex platform requires less technique and training for use
  • Ergonomically designed with special indentions on each side for finger placement
  • Needle point is hidden before use and automatically retracts after use to avoid accidental needlestick injuries and cross-infection
  • Rendered inoperable after use, providing highest level of safety for healthcare workers and patients and meeting OSHA standards
  • Visual lockout feature clearly indicates that device has been used and is ready for disposal in a sharps container
  • High-flow volume suitable for cholesterol, hemoglobin and cardiac enzyme testing

US Patents 5,487,748; 6,258,112; 6,764,496

Specifications

Disposable Single-use
Depth (Metric) 1.8mm
Needle Gauge 28g
Color Lavendar
Quantity 1000 Case
Volume (English) 0.0001 to 0.001 oz.
Product Type Unistik™ 3 Safety Lancet
For Use With (Application) For ward blood glucose monitoring, HbA1c
Depth (English) 0.07 in.
Volume (Metric) 5 to 30μL