Learn More
Owen Mumford™ Unistik™ 3 Safety Lancets
Feature unique Comfort Zone™ technology that allows ultimate control and comfort when lancing
Supplier:  Owen Mumford™ AT 1044
 View more versions of this product
Catalog No. 2202931
Only null left
Description
Description
  • Preset and ready for use; simply twist off protective sterile cap and take sample
  • Needle point is hidden before use and automatically retracts after use to avoid accidental needlestick injuries and cross-infection
  • Have an additional visual lockout feature; should an attempt be made to misuse the device, there is a clear indication to the operator that the device has been used and should be disposed of in a suitable sharps container
Specifications
Chemical Identifiers
0.07 in.
Unistik™ 3 Safety Lancet
Sterile cap
1.8mm
Single-use
Specifications
Single-use
For ward blood glucose monitoring, cholesterol, HbA1c
200 Pack
5 to 30μL
Lavendar
1.8mm
Sterile cap
0.00016 to 0.00101 oz.
0.07 in.
28g
Unistik™ 3 Safety Lancet
Product Suggestions
Product Suggestions