missing translation for 'onlineSavingsMsg'
Learn More

Eagle™ Overpack Poly Drum with Screw-on Lid, 65 gal.
GREENER_CHOICE

Catalog No. 19338400
Change view
Click to view available options
Capacity (English):
65 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
19338400 65 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19338400 Supplier Eagle™ Supplier No. 1661

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Drum is constructed of yellow blow-molded high-density polyethylene (HDPE) with UV inhibitors, is durable, lightweight, and weather/chemical resistant. Includes a yellow screw-on lid and is ideal for safe storage, handling and transportation of hazardous materials, or bio waste disposal.

  • For use with larger non-damaged containers, cans, and drums of non-leaking hazardous materials
  • If used with leaking containers, must pack with sufficient sorbents
  • No free liquids

Specifications

Product Type Overpack Drum
Closure Included Yes
Closure Type Screw-on Lid
Capacity (English) 65 gal.
Capacity (Metric) 246.35 L
Color Yellow
For Use With (Application) Safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration
Material HDPE
Height (English) 33.75 in.
Height (Metric) 85.8 cm
Weight (English) 45 lb.
Show More Show Less