Learn More
Selleck Chemical Llc Birabresib (OTX015) 2mg 202590-98-5 MK 8628 DFS Item
Supplier:  Selleck Chemicals Co Ltd S73602MG
Birabresib (OTX015, MK 8628) is a potent BET bromodomain inhibitor with EC50 ranging from 10 to 19 nM for BRD2, BRD3, and BRD4 in cell-free assays. Birabresib inhibits the expression of Nuclear receptor binding SET domain protein 3 (NSD3) target genes. *For Research & Development use only. Product is not intended for drug, household, or human consumption.
Catalog No. NC1075402
Only null left
Explore available promotions