Learn More
Waters Limited Ostro Protein Precipitation & Phospholipid Removal Plate, 25 mg, 1/pk DFS Item
Supplier:  Waters Limited 186005518
Ostro sample preparation products provide a novel solution for the cleanup of phospholipids in plasma and serum. Requiring minimal to no method development and using a simple protocol, the technology can be quickly implemented in order to optimize your laboratory's work flow. Providing cleaner, more reproducible extracts than competitive phospholipid devices or techniques, Ostro allows for more sensitive analyses, increased sample throughput, and reduced instrument downtime.
Catalog No. NC0022768
Only null left
Explore available promotions