missing translation for 'onlineSavingsMsg'
Learn More

Manganese Chloride, Tetrahydrate, USP, 98-101%, Spectrum™ Chemical

Catalog No. 18604186
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604186 12 kg Fiber Drum
18604183 125 g Fiber Drum
18604184 500 g Fiber Drum
18604185 2.5 kg Fiber Drum
18604187 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18604186 Supplier Spectrum Chemical Mfg Cor Supplier No. M110612KG
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

M1106, 13446-34-9, MnCl2·4H2O

Spectrum™ Chemical Manganese Chloride, Tetrahydrate, USP is indicated for use as a supplement to intravenous solutions given for total parenteral nutrition (TPN) to maintain Manganese serum levels. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 13446-34-9
Molecular Formula Cl2H8MnO4
Molecular Weight (g/mol) 197.90
MDL Number MFCD00149792
InChI Key CNFDGXZLMLFIJV-UHFFFAOYSA-L
IUPAC Name manganese(2+) tetrahydrate dichloride
SMILES O.O.O.O.[Cl-].[Cl-].[Mn++]

Specifications

Physical Form Fine Crystals or Crystalline Powder
CAS Max % 100%
Quantity 12 kg
Loss on Drying 36 to 38.5%
Packaging Fiber Drum
Percent Purity 98 to 101%
Grade USP
Chemical Name or Material Manganese Chloride, Tetrahydrate