missing translation for 'onlineSavingsMsg'
Learn More

Osthole 98.0+%, TCI America™

Catalog No. O0426250MG
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O0426250MG 250 mg
O04261G 1 g
O04265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. O0426250MG Supplier TCI America Supplier No. O0426250MG
Only null left

Chemical Identifiers

CAS 484-12-8
Molecular Formula C15H16O3
Molecular Weight (g/mol) 244.29
MDL Number MFCD00076049
InChI Key MBRLOUHOWLUMFF-UHFFFAOYSA-N
Synonym 7-Methoxy-8-(3-methyl-2-butenyl)coumarin
PubChem CID 10228
ChEBI CHEBI:69832
IUPAC Name 7-methoxy-8-(3-methylbut-2-enyl)chromen-2-one
SMILES CC(=CCC1=C(C=CC2=C1OC(=O)C=C2)OC)C

Specifications

Melting Point 84°C
Color White-Yellow
Quantity 250 mg
Formula Weight 244.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Osthole
RTECSNumber GN7700000
TSCA No