Learn More
ACROBiosystems Inc Recombinant Protein;Human Osteopontin (OPN) Protein;HEK293;100UG;OPN,SPP1,BNSP,OPN,Uropontin,Nephropontin,Osteopontin,SPP-1,BSP-1,ETA-1,BSPI,BSP1,ETA1
Numéro de catalogue. 501658039
Encompass
Recombinant Protein;Human Osteopontin (OPN) Protein;100UG;Human OPN, His Tag (OPN-H5227) is expressed from human 293 cells (HEK293). It contains AA Ile 17 - Asn 300 (Accession # AAH07016).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 33 kDa. The protein migrates as 60-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Numéro de catalogue. 501658039 Fournisseur ACROBiosystems Inc Code fournisseur OPNH5227100UG
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Recombinant Protein;Human Osteopontin (OPN) Protein;100UG;Human OPN, His Tag (OPN-H5227) is expressed from human 293 cells (HEK293). It contains AA Ile 17 - Asn 300 (Accession # AAH07016).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 33 kDa. The protein migrates as 60-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.