missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Osmosis Kit

Catalog No. S23637
Change view
Click to view available options
Class Size:
1 Lab Group
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S23637 1 Lab Group
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S23637 Supplier Eisco™ Supplier No. BI0204

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed for demonstration of osmotic pressure at an elementary level in biology. Osmosis Kit is ideal for students to understand the concept of Osmosis.

  • Semi-permeable attached to wide end of funnel having an indicator tube
  • Funnel is fixed in reservoir
  • Supplied with teat dropper pipette

Specifications

Grade 7 to 12
Includes Semipermeable membrane, rubber bands, thistle tube, reservoir, and teat dropper pipette
Class Size 1 Lab Group
Product Type Osmosis Kit