missing translation for 'onlineSavingsMsg'
Learn More

Osmium(III) chloride trihydrate, Premion™, 99.99% (metals basis), Os 52-56%

Catalog No. AA3348001
Change view
Click to view available options
Quantity:
0.25 g
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3348001 0.25 g
AA3348003 1 g
AA3348006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3348001 Supplier Thermo Scientific Chemicals Supplier No. 03348001

CAS: 135296-80-9 | Cl3H6O3Os | 350.625 g/mol

Osmium(III) chloride trihydrate is used as a reagent for preparation of dichlorodihydridoosmium complexes. It is also useful as starting materials and an efficient alkane oxidation catalyst in the presence of pyridine and hydrogen oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 135296-80-9
Molecular Formula Cl3H6O3Os
Molecular Weight (g/mol) 350.625
MDL Number MFCD00011148
InChI Key UACQLNPCDXDCID-UHFFFAOYSA-K
Synonym osmium iii chloride trihydrate, osmium trichloride trihydrate, cl3os.3h2o, acmc-20ak04, trichloroosmium-water 1/3, trichloroosmium trihydrate, osmium iii chloride trihydrate, premion
PubChem CID 57348105
IUPAC Name trichloroosmium;trihydrate
SMILES O.O.O.Cl[Os](Cl)Cl

Specifications

Melting Point (decomposition)
Color Black-Red
Physical Form Crystalline
Quantity 0.25 g
Linear Formula OsCl3·3H2O
UN Number UN3260
Solubility Information Soluble in water.
Formula Weight 350.60 (296.56 Anhydrous)
Percent Purity 99.99%
Grade Premion
Concentration or Composition (by Analyte or Components) Os 52 to 56%
Assay (metals basis)
Chemical Name or Material Osmium(III) chloride trihydrate
Show More Show Less
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H301-H311-H314-H318-H332
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 236-587-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only