missing translation for 'onlineSavingsMsg'
Learn More

Osmium(III) chloride hydrate

CAS: 14996-60-2 | Cl3H2OOs | 314.60 g/mol

$370.67 - $370.67

Chemical Identifiers

CAS 14996-60-2
Molecular Formula Cl3H2OOs
Molecular Weight (g/mol) 314.60
MDL Number MFCD00149815
InChI Key KHHOLOMEVBPRQY-UHFFFAOYSA-K
Synonym osmium iii chloride hydrate, osmium chloride oscl3 , hydrate 8ci,9ci, trichloroosmium hydrate, cl3os.h2o, trichloroosmium-water 1/1
PubChem CID 16211485
SMILES O.Cl[Os](Cl)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC318705000
SDS View Documents
Thermo Scientific Chemicals
318705000
500 mg Glass bottle
Each for $370.67
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

14996-60-2
314.60
KHHOLOMEVBPRQY-UHFFFAOYSA-K
16211485
Cl3H2OOs
MFCD00149815
osmium iii chloride hydrate, osmium chloride oscl3 , hydrate 8ci,9ci, trichloroosmium hydrate, cl3os.h2o, trichloroosmium-water 1/1
O.Cl[Os](Cl)Cl

Specifications

>500.0°C
14996-60-2
500 mg
OsCl3·xH2O
osmium iii chloride hydrate, osmium chloride oscl3 , hydrate 8ci,9ci, trichloroosmium hydrate, cl3os.h2o, trichloroosmium-water 1/1
KHHOLOMEVBPRQY-UHFFFAOYSA-K
trichloroosmium;hydrate
16211485
Glass bottle
Black to Red
Crystals or Powder
Cl3H2OOs
MFCD00149815
Solubility in water: soluble
O.Cl[Os](Cl)Cl
314.60
296.56
Osmium(III) chloride hydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.

GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for s

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only