missing translation for 'onlineSavingsMsg'
Learn More

Osmium(III) chloride hydrate

Catalog No. AC318705000
Change view
Click to view available options
Quantity:
500 mg
2.5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC318705000 500 mg Glass bottle
AC318700025 2.5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC318705000 Supplier Thermo Scientific Chemicals Supplier No. 318705000
Only null left

CAS: 14996-60-2 | Cl3H2OOs | 314.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14996-60-2
Molecular Formula Cl3H2OOs
Molecular Weight (g/mol) 314.60
MDL Number MFCD00149815
InChI Key KHHOLOMEVBPRQY-UHFFFAOYSA-K
Synonym osmium iii chloride hydrate, osmium chloride oscl3 , hydrate 8ci,9ci, trichloroosmium hydrate, cl3os.h2o, trichloroosmium-water 1/1
PubChem CID 16211485
SMILES O.Cl[Os](Cl)Cl

Specifications

Melting Point >500.0°C
Color Black to Red
Physical Form Crystals or Powder
Quantity 500 mg
Linear Formula OsCl3·xH2O
Solubility Information Solubility in water: soluble
IUPAC Name trichloroosmium;hydrate
Formula Weight 296.56
Packaging Glass bottle
Chemical Name or Material Osmium(III) chloride hydrate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for s

RUO – Research Use Only