missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Osmium Catalyst supported on Magnetite (0.07-0.09mmol/g), TCI America™
Specifications
Specifications
| Color | Black |
| Quantity | 1 g |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Osmium Catalyst supported on Magnetite (0.07-0.09mmol/g) |
Safety and Handling
| TSCA | No |