Learn More
QuidelOrtho Reagent Red Cells
Reagent red blood cells are used in the detection and identification of a variety of blood group antibodies
$246.99 - $481.38
Specifications
For Use With (Application) For detection and identification of a variety of Blood group Antibodies
View More Specs
Alert:
All blood products should be treated as potentially infectious. Source material from which this product was derived was found negative when tested in accordance with current FDA required tests. No known test methods can offer assurance that products derived from human blood will not transmit infectious agents.
Products 2
Catalog Number Mfr. No. Quantity Type Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Type Price Quantity & Availability  
23287330
Encompass
View Documents Promotion Details
QuidelOrtho
719810
1 x 10 mL Coombs Control Reagent Red Cells
Each for $246.99
Only null left
23287501
Encompass
View Documents Promotion Details
QuidelOrtho
719201
2 x 10 mL Affirmagen™
Pack for $481.38
Only null left
Description
Description
  • Tests include hemagglutination and hemolysis-based results
  • A variety of testing functions are available, including:
    • Detection and/or identification of unexpected blood group antibodies
    • Affirmation of expected group antibodies
    • Serum grouping
    • Confirmation of negative antiglobulin tests
  • Screening for RhD (Rh0) positive fetal red blood cells
Specifications
Specifications
For detection and identification of a variety of Blood group Antibodies