missing translation for 'onlineSavingsMsg'
Learn More

Ortho-Phosphoric Acid, MilliporeSigma

85%, for analysis EMSURE™ ACS,ISO,Reag. Ph Eur

$131.21 - $337.49

Chemical Identifiers

CAS 7664-38-2
Molecular Weight (g/mol) 98 g/mol
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M1005731000
View Documents
Technidata
1005731000
1 L
Each for $171.86
Only null left
 
M1005732500
View Documents
Technidata
1005732500
2.5 L
Each for $337.49
Only null left
 
M1005730510
View Documents
Technidata
1005730510
0.5 L
Each for $131.21
Only null left
 
Description

Description

Concentrated phosphoric acid is approx. 85% H3PO4 (aqueous)

Specifications

Chemical Identifiers

7664-38-2
98 g/mol

Specifications

7664-38-2
1 L
85%
Ortho-Phosphoric acid
Plastic bottle
98 g/mol
ACS
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'