missing translation for 'onlineSavingsMsg'
Learn More

Ortho-Phosphoric Acid, MilliporeSigma

Catalog No. M1005732500
Change view
Click to view available options
Quantity:
0.5 L
1 L
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1005732500 2.5 L
M1005731000 1 L
M1005730510 0.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1005732500 Supplier Technidata Supplier No. 1.00573.2500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

85%, for analysis EMSURE™ ACS,ISO,Reag. Ph Eur

Concentrated phosphoric acid is approx. 85% H3PO4 (aqueous)

Chemical Identifiers

CAS 7664-38-2
Molecular Weight (g/mol) 98 g/mol

Specifications

Packaging Plastic bottle
Quantity 2.5 L
Percent Purity 85%
Grade ACS
Chemical Name or Material Ortho-Phosphoric acid