missing translation for 'onlineSavingsMsg'
Learn More

ortho-Phosphoric acid 85%, Suprapur™, MilliporeSigma™

Catalog No. M1005522500
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
2.5 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1005522500 2.5 L
M1005520250 250 mL
M1005521000 1 L
M1005520500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. M1005522500 Supplier Technidata Supplier No. 1.00552.2500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7664-38-2
Molecular Formula H3O4P
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
IUPAC Name phosphoric acid

Specifications

pH < 0.5 (100 g/L, H2O, 20°C)
Melting Point 21°C
Boiling Point 158°C (1013 mbar)
Density 1.71 g/cm3 (at 20°C)
Packaging Plastic Bottle
Vapor Pressure 2 hPa (20°C)
Assay Percent Range ≥ 85.0%
Quantity 2.5 L
Viscosity 30.5 mm2/s (20°C)
Synonym o-Phosphoric acid, Orthophosphoric acid
SMILES OP(O)(O)=O
Molecular Weight (g/mol) 97.99
Show More Show Less
Recommended Storage Store above 15°C