missing translation for 'onlineSavingsMsg'
Learn More

ortho-Phosphoric acid 85%, Suprapur™, MilliporeSigma™

Catalog No. M1005520250
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
2.5 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1005520250 250 mL
M1005521000 1 L
M1005520500 500 mL
M1005522500 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. M1005520250 Supplier Technidata Supplier No. 1.00552.0250

Chemical Identifiers

CAS 7664-38-2
Molecular Formula H3O4P
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
PubChem CID 1004
ChEBI CHEBI:26078
IUPAC Name phosphoric acid

Specifications

Color Colorless
Density 1.71g/cm3 (20°C)
Packaging Plastic Bottle
Vapor Pressure 2 hPa (20°C)
Assay Percent Range 85%
Quantity 250 mL
Physical Form Liquid
Synonym orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
Solubility Information Soluble (2°C)
SMILES OP(=O)(O)O
Molecular Weight (g/mol) 97.994
Grade Suprapur
Chemical Name or Material Suprapur™
Show More Show Less
Hazard Category SignalWord: Danger
Recommended Storage Store above +15°C