missing translation for 'onlineSavingsMsg'
Learn More
Learn More
QuidelOrtho Workstation
Simple-to-use combined centrifuge and incubator with ID-MTS™ Gel Technology
$25,795.77
| Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
OK6904630
|
QuidelOrtho
6904630 |
Workstation |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Precision and accuracy
- Uses proven reliable ID-MTS™ Gel Cards
- Validated system of ID-MTS™ Gel Card with centrifuge and incubator
- Clear, stable, well-defined end points
- Objective interpretation of results and full assay portfolio
Convenient, compact 2 in 1 QuidelOrtho Workstation
- Integrated system of ID-MTS™ Gel Cards, centrifuge and incubator
- Light weight and small footprint for space-constrained labs
- Labs can standardize on one technology
Improved Process Controls
- Incubator temperature monitoring with visual indicator
- Unique beeping pattern to indicate cycle completion
- Enhanced error code intelligence for troubleshooting
- Quiet centrifuge with translucent panels for visual monitoring
Ease of Use
- Language-neutral icons and enhanced alarm systems
- Easy to perform centrifuge QC using a tachometer
- Quick access to holding area for centrifuge balance-card
Designed with the environment in mind
- Reduction in device weight by 33%
- Reduction in external packaging by 54%
- Reduction in Reference Guide size by 22%
- Reduced energy consumption by 70%
Applications:
- ABO Grouping/Rh Testing
- NeoNatal Blood Type (Forward Only)
- Blood Unit Confirmation (Group A, B, AB)
- Blood Unit Confirmation (Group O)
- Indirect Antiglobulin Testing
- Compatibility Testing (Coombs Crossmatch and Immediate Spin Crossmatch)
- Direct Antiglobulin Testing
- Rh Phenotyping
- Rh Typing