missing translation for 'onlineSavingsMsg'
Learn More

Orthanilic acid, 95%

Catalog No. AAH3176322
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3176322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3176322 Supplier Thermo Scientific Chemicals Supplier No. H3176322
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 88-21-1 | C6H7NO3S | 173.186 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

CAS 88-21-1
Melting Point >300°C
Odor Odorless
Molecular Formula C6H7NO3S
Quantity 100 g
MDL Number MFCD00007705
Synonym orthanilic acid, aniline-2-sulfonic acid, o-aminobenzenesulfonic acid, o-sulfanilic acid, aniline-o-sulfonic acid, 2-sulfanilic acid, aniline-o-sulphonic acid, 1-aminobenzene-2-sulfonic acid, 2-aminobenzenesulphonic acid, benzenesulfonic acid, 2-amino
InChI Key ZMCHBSMFKQYNKA-UHFFFAOYSA-N
SMILES C1=CC=C(C(=C1)N)S(=O)(=O)O
IUPAC Name 2-aminobenzenesulfonic acid
Molecular Weight (g/mol) 173.186
PubChem CID 6926
ChEBI CHEBI:1015
Formula Weight 173.19
Percent Purity 97%
Chemical Name or Material Orthanilic acid
Show More Show Less
Hazard Category H314-H317
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P272-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 201-810-9
RTECSNumber DB4727700
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only