missing translation for 'onlineSavingsMsg'
Learn More

Orotic acid hydrate, 98%

Catalog No. AAA1859430
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1859430 250 g
AAA1859418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1859430 Supplier Thermo Scientific Chemicals Supplier No. A1859430
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 50887-69-9 | C5H4N2O4 | 156.10 g/mol

Orotic acid hydrate is used in cosmetic and food industry. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Orotic acid hydrate is used in cosmetic and food industry. It is also used as a pharmaceutical intermediate.

Solubility
Soluble in water (1.7 mg/ml), and ammonium hydroxide. Insoluble in chloroform.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 50887-69-9
Molecular Formula C5H4N2O4
Molecular Weight (g/mol) 156.10
MDL Number MFCD00006027
InChI Key PXQPEWDEAKTCGB-UHFFFAOYSA-N
Synonym orotic acid monohydrate, orotic acid hydrate, 2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid hydrate, lactinium, oroticum acidum, 2,6-dioxo-1,2,3,6-tetrahydro-4-pyrimidinecarboxylic acid, orotc acd hydrate?, uracil-6-carboxylic acid monohydrate, uracil-4-carboxylic acid hydrate
PubChem CID 16218504
IUPAC Name 2,4-dioxo-1H-pyrimidine-6-carboxylic acid;hydrate
SMILES OC(=O)C1=CC(=O)NC(=O)N1

Specifications

Melting Point >300°C
Quantity 250 g
Beilstein 383901
Merck Index 14,6876
Solubility Information Soluble in water (1.7mg/ml),and ammonium hydroxide. Insoluble in chloroform.
Formula Weight 156.10 (Anhydrous)
Percent Purity 98%
Chemical Name or Material Orotic acid hydrate
Hazard Category H302-H315-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 200-619-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only